Assay Details
Gene Symbol: agxtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213162.1 | NP_998327.1 | 7 - 8 | 882 | 110 | |
GenBank mRNA | BC056520.1 | - | 7 - 8 | 882 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
