Assay Details
Gene Symbol: ssr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001399.2 | NP_001001399.2 | 2 - 3 | 254 | 99 | |
GenBank mRNA | AF364084.1 | - | 1 - 2 | 150 | 99 | |
BC075915.1 | - | 2 - 3 | 178 | 99 | ||
BC153439.1 | - | 680 | 99 | |||
EH445458.1 | - | 2 - 3 | 257 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
