Assay Details
Gene Symbol: shoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044786.1 | NP_001038251.1 | 4 - 5 | 1353 | 82 | |
XM_005161796.4 | XP_005161853.1 | 4 - 5 | 1354 | 82 | ||
GenBank mRNA | BC155579.1 | - | 4 - 5 | 1329 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
