Assay Details
Gene Symbol: heca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045126.2 | NP_001038591.1 | 3 - 4 | 1509 | 99 | |
GenBank mRNA | BC162405.1 | - | 3 - 4 | 1509 | 99 | |
BC162665.1 | - | 3 - 4 | 1509 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
