Assay Details
Gene Symbol: pgghg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077725.1 | NP_001071193.1 | 6 - 7 | 1232 | 116 | |
GenBank mRNA | BC125951.1 | - | 6 - 7 | 1232 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
