Assay Details
Gene Symbol: nadkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080002.1 | NP_001073471.1 | 4 - 5 | 511 | 131 | |
XM_009298640.3 | XP_009296915.1 | 2 - 3 | 321 | 131 | ||
GenBank mRNA | BC125853.1 | - | 4 - 5 | 511 | 131 | |
BC134965.1 | - | 4 - 5 | 534 | 131 | ||
BC165350.1 | - | 3 - 4 | 500 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
