Assay Details
Gene Symbol: myct1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082917.1 | NP_001076386.1 | 1 - 2 | 325 | 104 | |
GenBank mRNA | BC152519.1 | - | 1 - 2 | 302 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
