Assay Details
Gene Symbol: rgs13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089512.1 | NP_001082981.1 | 3 - 4 | 239 | 122 | |
GenBank mRNA | BC134221.1 | - | 3 - 4 | 239 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
