Assay Details
Gene Symbol: abr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100016.1 | NP_001093486.1 | 4 - 5 | 496 | 84 | |
NM_001368694.1 | NP_001355623.1 | 19 - 20 | 2993 | 84 | ||
XM_003198714.5 | XP_003198762.4 | 19 - 20 | 2375 | 84 | ||
XM_005168629.4 | XP_005168686.1 | 4 - 5 | 800 | 84 | ||
XM_005172023.4 | XP_005172080.3 | 19 - 20 | 2058 | 84 | ||
XM_017356374.2 | XP_017211863.2 | 14 - 15 | 1499 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
