Assay Details
Gene Symbol: abr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100016.1 | NP_001093486.1 | 5 - 6 | 623 | 72 | |
NM_001368694.1 | NP_001355623.1 | 20 - 21 | 3120 | 72 | ||
XM_003198714.5 | XP_003198762.4 | 20 - 21 | 2502 | 72 | ||
XM_005168629.4 | XP_005168686.1 | 5 - 6 | 927 | 72 | ||
XM_005172023.4 | XP_005172080.3 | 20 - 21 | 2185 | 72 | ||
XM_017356374.2 | XP_017211863.2 | 15 - 16 | 1626 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
