Assay Details
Gene Symbol: trioa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104526.1 | NP_001097996.1 | 46 - 47 | 6686 | 72 | |
XM_009294421.3 | XP_009292696.1 | 46 - 47 | 6865 | 72 | ||
XM_021468075.1 | XP_021323750.1 | 47 - 48 | 6958 | 72 | ||
XM_021468076.1 | XP_021323751.1 | 47 - 48 | 6959 | 72 | ||
XM_021468077.1 | XP_021323752.1 | 47 - 48 | 6959 | 72 | ||
XM_021468078.1 | XP_021323753.1 | 47 - 48 | 6960 | 72 | ||
XM_021468079.1 | XP_021323754.1 | 47 - 48 | 6961 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
