Assay Details
Gene Symbol: mkrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152979.2 | NP_694511.1 | 3 - 4 | 418 | 141 | |
XM_005155668.4 | XP_005155725.1 | 3 - 4 | 436 | 141 | ||
GenBank mRNA | AF277172.1 | - | 3 - 4 | 367 | 141 | |
BC045986.1 | - | 3 - 4 | 371 | 141 | ||
BC065352.1 | - | 3 - 4 | 362 | 141 | ||
CN321915.1 | - | 3 - 4 | 359 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
