Assay Details
Gene Symbol: paics | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199537.2 | NP_955831.1 | 7 - 8 | 1118 | 64 | |
GenBank mRNA | BC048051.1 | - | 7 - 8 | 1081 | 64 | |
BC071527.1 | - | 7 - 8 | 1118 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
