Assay Details
Gene Symbol: sec22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199698.1 | NP_955992.1 | 3 - 4 | 489 | 70 | |
XM_021478609.1 | XP_021334284.1 | 3 - 4 | 70 | |||
GenBank mRNA | BC049530.1 | - | 3 - 4 | 489 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
