Assay Details
Gene Symbol: gps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200973.3 | NP_957267.1 | 8 - 9 | 1295 | 85 | |
XM_005165131.3 | XP_005165188.1 | 8 - 9 | 916 | 85 | ||
XM_017356047.2 | XP_017211536.1 | 8 - 9 | 1157 | 85 | ||
XM_021476040.1 | XP_021331715.1 | 9 - 10 | 929 | 85 | ||
GenBank mRNA | BC096776.1 | - | 8 - 9 | 1296 | 85 | |
BC164184.1 | - | 7 - 8 | 772 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
