Assay Details
Gene Symbol: mydgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002480.1 | NP_001002480.1 | 6 | 1344 | 137 | |
GenBank mRNA | BC076331.1 | - | 6 | 1344 | 137 |
Gene Symbol: LOC101884287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017353436.2 | XP_017208925.1 | 2 | 5724 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
