Assay Details
Gene Symbol: f2rl1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098778.1 | NP_001092248.1 | 2 | 1249 | 134 | |
XM_005161489.4 | XP_005161546.1 | 2 | 1337 | 134 | ||
GenBank mRNA | BC142840.1 | - | 2 | 1249 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
