Assay Details
Gene Symbol: cela1.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003737.1 | NP_001003737.1 | 7 - 8 | 797 | 138 | |
GenBank mRNA | BC078376.1 | - | 7 - 8 | 797 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
