Assay Details
Gene Symbol: gna14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003753.1 | NP_001003753.1 | 4 - 5 | 641 | 90 | |
GenBank mRNA | BC077106.1 | - | 4 - 5 | 641 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
