Assay Details
Gene Symbol: gna14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003753.1 | NP_001003753.1 | 6 - 7 | 917 | 125 | |
GenBank mRNA | BC077106.1 | - | 6 - 7 | 917 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
