Assay Details
Gene Symbol: nop14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003831.1 | NP_001003831.1 | 1854 | 72 | ||
GenBank mRNA | AY648711.1 | - | 1854 | 72 | ||
BC085643.1 | - | 1850 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
