Assay Details
Gene Symbol: rtkn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005937.2 | NP_001005937.2 | 7 - 8 | 897 | 68 | |
GenBank mRNA | BC083519.1 | - | 7 - 8 | 897 | 68 | |
BC092947.1 | - | 7 - 8 | 898 | 68 | ||
BC098873.1 | - | 7 - 8 | 897 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
