Assay Details
Gene Symbol: kansl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005990.1 | NP_001005990.1 | 6 - 7 | 926 | 74 | |
XM_005162163.4 | XP_005162220.1 | 6 - 7 | 1014 | 74 | ||
GenBank mRNA | BC083416.1 | - | 6 - 7 | 926 | 74 | |
BC155307.1 | - | 6 - 7 | 902 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
