Assay Details
Gene Symbol: pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008628.1 | NP_001008628.1 | 3 - 4 | 852 | 118 | |
GenBank mRNA | BC086729.1 | - | 3 - 4 | 852 | 118 | |
BC165767.1 | - | 3 - 4 | 774 | 118 | ||
BC169189.1 | - | 835 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
