Assay Details
Gene Symbol: grtp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017810.1 | NP_001017810.1 | 2 - 3 | 227 | 127 | |
GenBank mRNA | BC093293.1 | - | 2 - 3 | 227 | 127 | |
BC164165.1 | - | 2 - 3 | 249 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
