Assay Details
Gene Symbol: timm10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017829.1 | NP_001017829.1 | 1 - 2 | 172 | 72 | |
GenBank mRNA | BC092787.1 | - | 1 - 2 | 172 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
