Assay Details
Gene Symbol: acot9.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020744.1 | NP_001018580.1 | 4 - 5 | 408 | 86 | |
XM_005162711.4 | XP_005162768.1 | 5 - 6 | 478 | 86 | ||
GenBank mRNA | BC095220.1 | - | 4 - 5 | 408 | 86 | |
BC159215.1 | - | 5 - 6 | 371 | 86 | ||
BC171360.1 | - | 5 - 6 | 426 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
