Assay Details
Gene Symbol: acot9.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020744.1 | NP_001018580.1 | 8 - 9 | 706 | 77 | |
GenBank mRNA | BC095220.1 | - | 8 - 9 | 706 | 77 | |
BC159215.1 | - | 9 - 10 | 669 | 77 | ||
BC171360.1 | - | 9 - 10 | 724 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
