Assay Details
Gene Symbol: mrpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024399.1 | NP_001019570.1 | 3 - 4 | 560 | 85 | |
GenBank mRNA | BC095131.1 | - | 3 - 4 | 560 | 85 | |
BC154265.1 | - | 3 - 4 | 511 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
