Assay Details
Gene Symbol: c1qtnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029953.1 | NP_001025124.1 | 2 - 3 | 337 | 82 | |
GenBank mRNA | BC098626.1 | - | 2 - 3 | 337 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
