Assay Details
Gene Symbol: gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105595.1 | NP_001099065.1 | 1 - 2 | 208 | 91 | |
GenBank mRNA | AF425745.1 | - | 1 - 2 | 231 | 91 | |
BC107607.1 | - | 1 - 2 | 255 | 91 | ||
BC153601.1 | - | 1 - 2 | 208 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
