Assay Details
Gene Symbol: mea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002111.1 | NP_001002111.1 | 2 - 3 | 369 | 87 | |
XM_005156496.4 | XP_005156553.1 | 2 - 3 | 406 | 87 | ||
GenBank mRNA | BC071483.1 | - | 2 - 3 | 369 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
