Assay Details
Gene Symbol: chrna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042684.2 | NP_001036149.1 | 4 - 5 | 518 | 106 | |
GenBank mRNA | BC171403.1 | - | 4 - 5 | 469 | 106 | |
BC171405.1 | - | 4 - 5 | 469 | 106 | ||
DQ822507.1 | - | 4 - 5 | 521 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
