Assay Details
Gene Symbol: snx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077368.2 | NP_001070836.1 | 3 - 4 | 540 | 79 | |
GenBank mRNA | BC124665.1 | - | 3 - 4 | 530 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
