Assay Details
Gene Symbol: stk35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077616.1 | NP_001071084.1 | 1 - 2 | 652 | 114 | |
GenBank mRNA | BC125863.1 | - | 1 - 2 | 652 | 114 | |
BC164504.1 | - | 1 - 2 | 538 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
