Assay Details
Gene Symbol: surf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080173.1 | NP_001073642.1 | 3 - 4 | 300 | 65 | |
GenBank mRNA | BC129348.1 | - | 3 - 4 | 300 | 65 | |
BC164262.1 | - | 3 - 4 | 304 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
