Assay Details
Gene Symbol: sgo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080620.1 | NP_001074089.1 | 4 - 5 | 694 | 133 | |
XM_021468141.1 | XP_021323816.1 | 4 - 5 | 792 | 133 | ||
GenBank mRNA | BC129193.1 | - | 4 - 5 | 694 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
