Assay Details
Gene Symbol: sgo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080620.1 | NP_001074089.1 | 9 - 10 | 1640 | 77 | |
XM_021468141.1 | XP_021323816.1 | 8 - 9 | 1567 | 77 | ||
GenBank mRNA | BC129193.1 | - | 9 - 10 | 1640 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
