Assay Details
Gene Symbol: etaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111157.1 | NP_001104627.1 | 3 - 4 | 424 | 110 | |
XM_009294369.2 | XP_009292644.1 | 4 - 5 | 722 | 110 | ||
GenBank mRNA | BC153950.1 | - | 3 - 4 | 424 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
