Assay Details
Gene Symbol: tnfrsf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113524.1 | NP_001106996.1 | 2 - 3 | 423 | 89 | |
XM_005166063.4 | XP_005166120.1 | 2 - 3 | 1279 | 89 | ||
GenBank mRNA | EU099311.1 | - | 2 - 3 | 423 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
