Assay Details
Gene Symbol: mrps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113608.1 | NP_001107080.1 | 2 - 3 | 210 | 96 | |
XM_021478290.1 | XP_021333965.1 | 2 - 3 | 205 | 96 | ||
GenBank mRNA | BC122242.1 | - | 2 - 3 | 210 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
