Assay Details
Gene Symbol: cox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114469.1 | NP_001107941.1 | 1 - 2 | 126 | 150 | |
XM_009291300.3 | XP_009289575.1 | 1 - 2 | 172 | 150 | ||
XM_021480952.1 | XP_021336627.1 | 1 - 2 | 172 | 150 | ||
GenBank mRNA | BC155287.1 | - | 1 - 2 | 126 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC155287.1, NM_001114469.1, XM_009291300.3, XM_021480952.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
