Assay Details
Gene Symbol: cox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114469.1 | NP_001107941.1 | 2 - 3 | 435 | 71 | |
XM_009291300.3 | XP_009289575.1 | 2 - 3 | 71 | |||
XM_021480952.1 | XP_021336627.1 | 2 - 3 | 71 | |||
GenBank mRNA | BC155287.1 | - | 2 - 3 | 435 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
