Assay Details
Gene Symbol: tmem30c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200596.2 | NP_956890.2 | 2 - 3 | 410 | 85 | |
GenBank mRNA | BC056764.1 | - | 2 - 3 | 410 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
