Assay Details
Gene Symbol: sap18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200720.2 | NP_957014.1 | 3 - 4 | 417 | 100 | |
GenBank mRNA | BC059482.1 | - | 3 - 4 | 355 | 100 | |
BC090729.1 | - | 2 - 3 | 1235 | 100 | ||
BC155197.1 | - | 3 - 4 | 365 | 100 | ||
BI709589.1 | - | 1 - 2 | 504 | 100 | ||
EH440312.1 | - | 3 - 4 | 352 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
