Assay Details
Gene Symbol: dnase2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114738.1 | NP_001108210.1 | 6 - 7 | 816 | 88 | |
XM_021466569.1 | XP_021322244.1 | 6 - 7 | 866 | 88 | ||
GenBank mRNA | BC159268.1 | - | 6 - 7 | 826 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
