Assay Details
Gene Symbol: ccl34a.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115069.1 | NP_001108541.1 | 2 - 3 | 209 | 107 | |
GenBank mRNA | AB331772.1 | - | 2 - 3 | 209 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
