Assay Details
Gene Symbol: mboat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122944.1 | NP_001116416.1 | 1 - 2 | 124 | 67 | |
GenBank mRNA | EU518498.1 | - | 1 - 2 | 124 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
