Assay Details
Gene Symbol: rab7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200928.1 | NP_957222.1 | 3 - 4 | 430 | 71 | |
GenBank mRNA | BC054602.1 | - | 3 - 4 | 430 | 71 | |
BC155202.1 | - | 3 - 4 | 414 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
