Assay Details
Gene Symbol: nme4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201195.1 | NP_957489.1 | 5 - 6 | 666 | 64 | |
GenBank mRNA | BC049030.1 | - | 5 - 6 | 666 | 64 | |
BC153640.1 | - | 4 - 5 | 494 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
